Assay Details
| Gene Symbol: Crmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136058.2 | NP_001129530.1 | 6 - 7 | 1046 | 71 | |
| NM_007765.4 | NP_031791.3 | 6 - 7 | 911 | 71 | ||
| GenBank mRNA | AB006714.1 | - | 6 - 7 | 883 | 71 | |
| AK158793.1 | - | 6 - 7 | 990 | 71 | ||
| AK159049.1 | - | 6 - 7 | 855 | 71 | ||
| BC031738.1 | - | 6 - 7 | 840 | 4505284 | 71 | |
| BC065046.1 | - | 6 - 7 | 1024 | 5686818 | 71 | |
| U72875.1 | - | 6 - 7 | 656 | 71 | ||
| Y09080.1 | - | 6 - 7 | 886 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crmp1
Genomic Map