Assay Details
| Gene Symbol: Gckr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144909.1 | NP_659158.1 | 5 - 6 | 467 | 106 | |
| GenBank mRNA | AK149377.1 | - | 5 - 6 | 494 | 106 | |
| BC012412.1 | - | 5 - 6 | 467 | 4159892 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gckr
Genomic Map