Assay Details
| Gene Symbol: Stx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289535.1 | NP_001276464.1 | 7 - 8 | 802 | 101 | |
| NM_001289536.1 | NP_001276465.1 | 6 - 7 | 724 | 101 | ||
| NM_026959.3 | NP_081235.2 | 7 - 8 | 802 | 101 | ||
| GenBank mRNA | AK007700.1 | - | 7 - 8 | 800 | 101 | |
| AK016910.1 | - | 7 - 8 | 744 | 101 | ||
| AK169273.1 | - | 7 - 8 | 746 | 101 | ||
| AK170165.1 | - | 7 - 8 | 798 | 101 | ||
| BC021362.1 | - | 6 - 7 | 714 | 5035350 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stx18
Genomic Map