Assay Details
| Gene Symbol: Epha5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007937.3 | NP_031963.2 | 2 - 3 | 1125 | 107 | |
| GenBank mRNA | AK034289.1 | - | 2 - 3 | 1124 | 107 | |
| AK049740.1 | - | 2 - 3 | 1126 | 107 | ||
| AK089130.1 | - | 2 - 3 | 520 | 107 | ||
| AK089143.1 | - | 2 - 3 | 984 | 107 | ||
| AK133698.1 | - | 2 - 3 | 1125 | 107 | ||
| BC057401.1 | - | 2 - 3 | 1103 | 5697758 | 107 | |
| U07357.1 | - | 2 - 3 | 665 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epha5
Genomic Map