Assay Details
| Gene Symbol: 4932443I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK030127.1 | - | 3 - 4 | 1576 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RAS p21 protein activator 3 | Rasa3 |
AI326412, C86362, E130011G04, GAPIII, scat |
-58.3kb | Anti-sense | Chr. 8 - 13567218 - 13677587 on NCBI Build GRCm38 |
| Downstream | CDC16 cell division cycle 16 | Cdc16 |
2700071J12Rik, 2810431D22Rik, APC6 |
17.6kb | Sense | Chr. 8 - 13757646 - 13781938 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4932443I19Rik
Genomic Map