Assay Details
Gene Symbol: Tmco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172282.2 | NP_758486.1 | 10 - 11 | 2137 | 75 | |
GenBank mRNA | AK033499.1 | - | 10 - 11 | 1898 | 75 | |
AK046065.1 | - | 10 - 11 | 1924 | 75 | ||
AK049974.1 | - | 10 - 11 | 1893 | 75 | ||
AK088866.1 | - | 10 - 11 | 1921 | 75 | ||
AK148252.1 | - | 10 - 11 | 1940 | 75 | ||
BC026890.1 | - | 3 - 4 | 410 | 75 | ||
BC039955.1 | - | 10 - 11 | 1883 | 4236297 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
