Assay Details
Gene Symbol: Tmco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172282.2 | NP_758486.1 | 11 - 12 | 2221 | 64 | |
GenBank mRNA | AK033499.1 | - | 11 - 12 | 1982 | 64 | |
AK046065.1 | - | 11 - 12 | 2008 | 64 | ||
AK049974.1 | - | 11 - 12 | 1977 | 64 | ||
AK088866.1 | - | 11 - 12 | 2005 | 64 | ||
AK148252.1 | - | 11 - 12 | 2024 | 64 | ||
BC026890.1 | - | 4 - 5 | 494 | 64 | ||
BC039955.1 | - | 11 - 12 | 1967 | 4236297 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
