Assay Details
Gene Symbol: Tmco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172282.2 | NP_758486.1 | 3 - 4 | 1071 | 107 | |
GenBank mRNA | AK033499.1 | - | 3 - 4 | 832 | 107 | |
AK046065.1 | - | 3 - 4 | 861 | 107 | ||
AK049974.1 | - | 3 - 4 | 830 | 107 | ||
AK083321.1 | - | 3 - 4 | 895 | 107 | ||
AK088866.1 | - | 3 - 4 | 855 | 107 | ||
AK148252.1 | - | 3 - 4 | 874 | 107 | ||
BC039955.1 | - | 3 - 4 | 817 | 4236297 | 107 | |
BC053487.1 | - | 3 - 4 | 846 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
