Assay Details
Gene Symbol: Tmco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172282.2 | NP_758486.1 | 4 - 5 | 1262 | 62 | |
GenBank mRNA | AK033499.1 | - | 4 - 5 | 1023 | 62 | |
AK046065.1 | - | 4 - 5 | 1052 | 62 | ||
AK049974.1 | - | 4 - 5 | 1021 | 62 | ||
AK083321.1 | - | 4 - 5 | 1086 | 62 | ||
AK088866.1 | - | 4 - 5 | 1046 | 62 | ||
AK148252.1 | - | 4 - 5 | 1065 | 62 | ||
BC039955.1 | - | 4 - 5 | 1008 | 4236297 | 62 | |
BC053487.1 | - | 4 - 5 | 1037 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
