Assay Details
| Gene Symbol: BB014433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037972.1 | 1 - 2 | 111 | 99 | ||
| GenBank mRNA | AK077224.1 | - | 1 - 2 | 103 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK077224.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P650333 | AK077224.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P650333 | NR_037972.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P650333 | AK077224.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P650333 | NR_037972.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156370 | BC139040.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156370 | AK077224.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156370 | NR_037972.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 361811 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808559 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 678323 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 168715 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124641 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P650333 | AK077224.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P650333 | NR_037972.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P650333 | AK077224.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P650333 | NR_037972.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156370 | BC139040.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156370 | AK077224.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156370 | NR_037972.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25678455 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124641 | - | - |
| Whole Mouse Genome G4122A | A_52_P650333 | AK077224.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P650333 | NR_037972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P650333 | AK077224.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P650333 | NR_037972.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113945 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124641 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06734352 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06734362 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07398028 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06734344 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06734350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25678472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07286447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07286442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06825621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07002522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07398028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07009105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06734344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07316219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06734355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07316217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07002518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07009096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07009091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25678444 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10577239 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10577238 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10577237 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800014196.mm.1 | NR_037972.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800014196.mm.1 | NR_037972.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800014196.mm.1 | TC0800001764.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800014194.mm.1 | NR_037972.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800014194.mm.1 | NR_037972.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800014194.mm.1 | TC0800001764.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800014195.mm.1 | NR_037972.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800014195.mm.1 | NR_037972.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800014195.mm.1 | TC0800001764.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800014199.mm.1 | NR_037972.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800014199.mm.1 | NR_037972.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800014199.mm.1 | TC0800001764.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 461234 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 103840452 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234722 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103840452 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103840452 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2926002 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234722 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2926002 | - | - |

BB014433
Genomic Map