Assay Details
| Gene Symbol: Cnga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007723.2 | NP_031749.2 | 1 - 2 | 246 | 119 | |
| GenBank mRNA | BI738168.1 | - | 1 - 2 | 246 | 119 | |
| CX564336.1 | - | 1 - 2 | 227 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_007723.2 transcripts.
Set Membership: |
|

Cnga1
Genomic Map