Assay Details
| Gene Symbol: Zcchc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019621.1 | - | 12 - 13 | 1419 | 90 | |
| BC107392.2 | - | 1344 | 40057902 | 90 | ||
| BC107393.2 | - | 1344 | 40057909 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zcchc4
Genomic Map