Assay Details
| Gene Symbol: Rab28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027295.2 | NP_081571.1 | 4 - 5 | 574 | 86 | |
| GenBank mRNA | AB232624.1 | - | 4 - 5 | 398 | 86 | |
| AK012286.1 | - | 4 - 5 | 577 | 86 | ||
| AK159493.1 | - | 4 - 5 | 575 | 86 | ||
| AK161391.1 | - | 4 - 5 | 455 | 86 | ||
| AK215070.1 | - | 3 - 4 | 310 | 86 | ||
| BC004580.1 | - | 4 - 5 | 574 | 3587008 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab28
Genomic Map