Assay Details
Gene Symbol: Cgref1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160149.1 | NP_001153621.1 | 3 - 4 | 208 | 93 | |
NM_026770.1 | NP_081046.1 | 3 - 4 | 292 | 93 | ||
GenBank mRNA | AK003420.1 | - | 3 - 4 | 219 | 93 | |
BC023116.1 | - | 3 - 4 | 292 | 4206019 | 93 | |
BQ555687.1 | - | 3 - 4 | 408 | 93 | ||
CB202031.1 | - | 3 - 4 | 209 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
