Assay Details
| Gene Symbol: Preb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294302.1 | NP_001281231.1 | 7 - 8 | 1220 | 62 | |
| GenBank mRNA | AK013330.1 | - | 7 - 8 | 1174 | 62 | |
| AK179426.1 | - | 2 - 3 | 176 | 62 | ||
More Information
Set Membership: |
|

Preb
Genomic Map