Assay Details
| Gene Symbol: Zbtb49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029162.2 | NP_083438.1 | 2 - 3 | 270 | 69 | |
| GenBank mRNA | AK043044.1 | - | 2 - 3 | 265 | 69 | |
| AK044991.1 | - | 2 - 3 | 297 | 69 | ||
| AK140546.1 | - | 2 - 3 | 265 | 69 | ||
| BC028988.1 | - | 2 - 3 | 255 | 5376301 | 69 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK148068.1 | - | 276 | Chr. - - [-] on Build | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC028988.1 transcripts.
Set Membership: |
|

Zbtb49
Genomic Map