Assay Details
| Gene Symbol: Mcph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042026.1 | - | 4 - 5 | 774 | 131 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK042026.1 transcripts.
Set Membership: |
|

Mcph1
Genomic Map