| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200003105.mm.1 |
NM_029862.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200003105.mm.1 |
NM_029862.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200003105.mm.1 |
TC0200000462.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200003096.mm.1 |
NM_001243239.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200003096.mm.1 |
NM_001243239.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200003096.mm.1 |
NM_001243240.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200003096.mm.1 |
NM_001243240.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200003096.mm.1 |
NM_029862.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200003096.mm.1 |
NM_029862.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200003096.mm.1 |
TC0200000462.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200003104.mm.1 |
NM_029862.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200003104.mm.1 |
NM_029862.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200003104.mm.1 |
TC0200000462.mm.1 |
MTA TC |