Assay Details
| Gene Symbol: Cacfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029862.4 | NP_084138.1 | 4 - 5 | 486 | 78 | |
| GenBank mRNA | AK039867.1 | - | 4 - 5 | 463 | 78 | |
| AK040379.1 | - | 4 - 5 | 393 | 78 | ||
| AK052562.1 | - | 4 - 5 | 447 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cacfd1
Genomic Map