Assay Details
| Gene Symbol: Slc5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019810.4 | NP_062784.3 | 7 - 8 | 905 | 141 | |
| GenBank mRNA | AF208031.1 | - | 7 - 8 | 905 | 141 | |
| AK033512.1 | - | 7 - 8 | 891 | 141 | ||
| AK085602.1 | - | 7 - 8 | 696 | 141 | ||
| BC003845.1 | - | 7 - 8 | 885 | 3497611 | 141 | |
| U27269.1 | - | 5 - 6 | 475 | 141 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc5a1
Genomic Map