Assay Details
| Gene Symbol: Mef2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133665.3 | NP_598426.1 | 9 - 10 | 1610 | 86 | |
| GenBank mRNA | AK155778.1 | - | 9 - 10 | 1611 | 86 | |
| BC011070.1 | - | 9 - 10 | 1578 | 3497896 | 86 | |
| S68893.1 | - | 10 - 11 | 1453 | 86 | ||
| S68895.1 | - | 9 - 10 | 1429 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mef2d
Genomic Map