Assay Details
| Gene Symbol: Galnt15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030166.3 | NP_084442.1 | 4 - 5 | 1465 | 144 | |
| GenBank mRNA | AB175682.1 | - | 4 - 5 | 1075 | 144 | |
| AK019470.1 | - | 4 - 5 | 1580 | 144 | ||
| AK030347.1 | - | 4 - 5 | 718 | 144 | ||
| AK050075.1 | - | 4 - 5 | 965 | 144 | ||
| AK090330.1 | - | 2 - 3 | 788 | 145 | ||
| AK163883.1 | - | 4 - 5 | 1341 | 144 | ||
| BC133711.1 | - | 4 - 5 | 1465 | 40143107 | 144 | |
More Information
Set Membership: |
|

Galnt15
Genomic Map