Assay Details
| Gene Symbol: Galnt15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030166.3 | NP_084442.1 | 7 - 8 | 1909 | 57 | |
| GenBank mRNA | AB175682.1 | - | 7 - 8 | 1519 | 57 | |
| AK019470.1 | - | 7 - 8 | 2024 | 57 | ||
| AK030347.1 | - | 7 - 8 | 1162 | 57 | ||
| AK050075.1 | - | 7 - 8 | 1409 | 57 | ||
| AK090330.1 | - | 5 - 6 | 1232 | 57 | ||
| AK163883.1 | - | 7 - 8 | 1785 | 57 | ||
| BC133711.1 | - | 7 - 8 | 1909 | 40143107 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt15
Genomic Map