Assay Details
| Gene Symbol: Galnt15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030166.3 | NP_084442.1 | 8 - 9 | 2013 | 80 | |
| GenBank mRNA | AK019470.1 | - | 8 - 9 | 2128 | 80 | |
| AK030347.1 | - | 8 - 9 | 1266 | 80 | ||
| AK050075.1 | - | 8 - 9 | 1513 | 80 | ||
| AK090330.1 | - | 6 - 7 | 1336 | 80 | ||
| AK163883.1 | - | 8 - 9 | 1889 | 80 | ||
| BC133711.1 | - | 8 - 9 | 2013 | 40143107 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt15
Genomic Map