Assay Details
| Gene Symbol: S100a7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199422.1 | NP_955454.1 | 2 - 3 | 263 | 62 | |
| GenBank mRNA | AY465109.1 | - | 2 - 3 | 263 | 62 | |
| AY582964.1 | - | 1 - 2 | 170 | 62 | ||
| BC107307.1 | - | 2 - 3 | 245 | 40053247 | 62 | |
| BC107308.1 | - | 2 - 3 | 245 | 40053248 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P63855 | NM_199422.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P63855 | AY465109.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P63855 | NM_199422.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P63855 | AY465109.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181480 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 246612 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179579 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P63855 | NM_199422.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P63855 | AY465109.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P63855 | NM_199422.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P63855 | AY465109.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326393 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326395 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326386 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22326368 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179579 | - | - |
| Whole Mouse Genome G4122A | A_52_P63855 | NM_199422.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P63855 | AY465109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P63855 | NM_199422.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P63855 | AY465109.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129410 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179579 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02576942 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02576948 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02478350 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02576947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22326401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02576942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02535643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02654266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02331623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02654269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02654268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02654244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02654264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326355 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03171615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02654257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03171623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02654246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02331624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02654253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02654271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02654249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02535647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02576948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02576947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22326410 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10493825 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493829 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493827 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493826 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493828 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493830 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493824 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300006316.mm.1 | NM_199422.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006316.mm.1 | NM_199422.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006316.mm.1 | TC0300000810.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006317.mm.1 | NM_199422.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006317.mm.1 | NM_199422.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006317.mm.1 | TC0300000810.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 536839 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1170100 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1170100 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001170100 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230080 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230080 | - | - |
| mouse-6 v1.1 (Illumina) | scl0381493.3_27-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1170100 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0381493.3_27-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1170100 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1230080 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1230080 | - | - |

S100a7a
Genomic Map