Assay Details
| Gene Symbol: Gfm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138591.2 | NP_613057.2 | 12 - 13 | 1635 | 87 | |
| GenBank mRNA | AF315511.1 | - | 12 - 13 | 1602 | 87 | |
| AK033214.1 | - | 12 - 13 | 1634 | 87 | ||
| BC031772.1 | - | 12 - 13 | 1602 | 4239980 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gfm1
Genomic Map