Assay Details
| Gene Symbol: Gfm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138591.2 | NP_613057.2 | 9 - 10 | 1345 | 68 | |
| GenBank mRNA | AK033214.1 | - | 9 - 10 | 1344 | 68 | |
| BC013093.1 | - | 9 - 10 | 1348 | 3584730 | 68 | |
| BC031772.1 | - | 9 - 10 | 1312 | 4239980 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gfm1
Genomic Map