Assay Details
Gene Symbol: Gatb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144896.4 | NP_659145.1 | 11 - 12 | 1424 | 123 | |
GenBank mRNA | AK028193.1 | - | 11 - 12 | 1423 | 123 | |
AK034702.1 | - | 11 - 12 | 1425 | 123 | ||
AK053978.1 | - | 11 - 12 | 1427 | 123 | ||
AK081800.1 | - | 11 - 12 | 1428 | 123 | ||
AK134831.1 | - | 11 - 12 | 1286 | 123 | ||
AK216487.1 | - | 2 - 3 | 199 | 123 | ||
BC005709.1 | - | 11 - 12 | 1426 | 3157973 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
