Assay Details
| Gene Symbol: Pmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025928.3 | NP_080204.1 | 2 - 3 | 271 | 61 | |
| GenBank mRNA | AF348510.1 | - | 2 - 3 | 281 | 61 | |
| AK011174.1 | - | 2 - 3 | 273 | 61 | ||
| AK012113.1 | - | 2 - 3 | 272 | 61 | ||
| AK013787.1 | - | 2 - 3 | 274 | 61 | ||
| AK078308.1 | - | 2 - 3 | 290 | 61 | ||
| AK150603.1 | - | 2 - 3 | 288 | 61 | ||
| AK168200.1 | - | 2 - 3 | 272 | 61 | ||
| BC034333.1 | - | 2 - 3 | 249 | 61 | ||
| BC037139.1 | - | 2 - 3 | 280 | 4952550 | 61 | |
| BC049223.1 | - | 2 - 3 | 270 | 3257320 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pmf1
Genomic Map