Assay Details
| Gene Symbol: Sema4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163489.1 | NP_001156961.1 | 7 - 8 | 747 | 67 | |
| NM_001163490.1 | NP_001156962.1 | 7 - 8 | 826 | 67 | ||
| NM_001163491.1 | NP_001156963.1 | 7 - 8 | 785 | 67 | ||
| NM_013658.3 | NP_038686.3 | 7 - 8 | 925 | 67 | ||
| GenBank mRNA | AK035918.1 | - | 6 - 7 | 787 | 67 | |
| AK077976.1 | - | 7 - 8 | 905 | 67 | ||
| AK152728.1 | - | 7 - 8 | 917 | 67 | ||
| AK153164.1 | - | 7 - 8 | 917 | 67 | ||
| AK154392.1 | - | 7 - 8 | 886 | 67 | ||
| BC025800.1 | - | 7 - 8 | 843 | 5102015 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sema4a
Genomic Map