Assay Details
| Gene Symbol: 1700025M24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040687.1 | 3 - 4 | 234 | 99 | ||
| GenBank mRNA | BC048655.1 | - | 3 - 4 | 231 | 99 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | CA465171.1 | - | 234 | Chr. - - [-] on Build | 99 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | furry homolog-like (Drosophila) | Fryl |
2010313D22Rik, 2310004H21Rik, 2510002A14Rik, 9030227G01Rik, mKIAA0826 |
-15.0kb | Sense | Chr. 5 - 73020191 - 73256645 on NCBI Build GRCm38 |
| Downstream | OCIA domain containing 1 | Ociad1 |
6030432N09Rik, AI481327, AW557942, Asrij, B230209J16Rik, BB021357, Emi2, Imi2, OCIA, TPA018 |
10.6kb | Anti-sense | Chr. 5 - 73292772 - 73314077 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700025M24Rik
Genomic Map