Assay Details
| Gene Symbol: Maea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021500.2 | NP_067475.2 | 1 - 2 | 77 | 75 | |
| GenBank mRNA | AK007307.1 | - | 1 - 2 | 48 | 75 | |
| AK034859.1 | - | 1 - 2 | 76 | 75 | ||
| AK049424.1 | - | 1 - 2 | 97 | 75 | ||
| AK145800.1 | - | 1 - 2 | 140 | 75 | ||
| AK152523.1 | - | 1 - 2 | 85 | 75 | ||
| AK168788.1 | - | 1 - 2 | 103 | 75 | ||
| AK169149.1 | - | 1 - 2 | 85 | 75 | ||
| AK218361.1 | - | 1 - 2 | 53 | 75 | ||
| BC039054.1 | - | 1 - 2 | 63 | 2648667 | 75 | |
| BC058687.1 | - | 1 - 2 | 75 | 6411707 | 75 | |
| CX212584.1 | - | 1 - 2 | 77 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007307.1, AK034859.1, BC039054.1 transcripts.
Set Membership: |
|

Maea
Genomic Map