Assay Details
| Gene Symbol: Maea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021500.2 | NP_067475.2 | 4 - 5 | 585 | 73 | |
| GenBank mRNA | AF263247.1 | - | 4 - 5 | 547 | 73 | |
| AK007307.1 | - | 4 - 5 | 556 | 73 | ||
| AK049424.1 | - | 4 - 5 | 605 | 73 | ||
| AK075637.1 | - | 3 - 4 | 425 | 73 | ||
| AK088854.1 | - | 4 - 5 | 593 | 73 | ||
| AK145800.1 | - | 4 - 5 | 648 | 73 | ||
| AK152523.1 | - | 4 - 5 | 593 | 73 | ||
| AK168788.1 | - | 4 - 5 | 611 | 73 | ||
| AK169149.1 | - | 4 - 5 | 593 | 73 | ||
| BC039054.1 | - | 4 - 5 | 571 | 2648667 | 73 | |
| BC058687.1 | - | 4 - 5 | 583 | 6411707 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Maea
Genomic Map