Assay Details
| Gene Symbol: Galt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK088379.1 | - | 2 - 3 | 293 | 94 | |
| BC010985.1 | - | 2 - 3 | 296 | 4038194 | 94 | |
| BY289084.1 | - | 2 - 3 | 297 | 94 | ||
| CX239267.1 | - | 2 - 3 | 289 | 94 | ||
| M96265.1 | - | 2 - 3 | 323 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Galt
Genomic Map