Assay Details
| Gene Symbol: Galt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK088379.1 | - | 4 - 5 | 422 | 93 | |
| BC010985.1 | - | 4 - 5 | 425 | 4038194 | 93 | |
| CX239267.1 | - | 4 - 5 | 418 | 93 | ||
| M96265.1 | - | 4 - 5 | 452 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Galt
Genomic Map