Assay Details
| Gene Symbol: Galt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK088379.1 | - | 6 - 7 | 605 | 101 | |
| BC010985.1 | - | 6 - 7 | 608 | 4038194 | 101 | |
| CX239267.1 | - | 6 - 7 | 601 | 101 | ||
| M96265.1 | - | 6 - 7 | 635 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Galt
Genomic Map