Assay Details
| Gene Symbol: Adam9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270996.1 | NP_001257925.1 | 11 - 12 | 1272 | 67 | |
| NM_007404.2 | NP_031430.2 | 11 - 12 | 1272 | 67 | ||
| GenBank mRNA | AK122188.1 | - | 11 - 12 | 1255 | 67 | |
| AK148174.1 | - | 11 - 12 | 1158 | 67 | ||
| AK155582.1 | - | 11 - 12 | 1233 | 67 | ||
| AK155672.1 | - | 11 - 12 | 1165 | 67 | ||
| AK161778.1 | - | 11 - 12 | 1196 | 67 | ||
| BC047156.1 | - | 11 - 12 | 1150 | 6488707 | 67 | |
| BC052710.1 | - | 11 - 12 | 1144 | 67 | ||
| U41765.1 | - | 11 - 12 | 1144 | 67 | ||
More Information
Set Membership: |
|

Adam9
Genomic Map