Assay Details
| Gene Symbol: Whsc1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK132725.1 | - | 7 - 8 | 1024 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Whsc1l1| Gene Symbol: Whsc1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK132725.1 | - | 7 - 8 | 1024 | 78 | |
Set Membership: |
|