Assay Details
| Gene Symbol: Whsc1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK156746.1 | - | 2 - 3 | 1149 | 78 | |
| AK170040.1 | - | 2 - 3 | 1212 | 78 | ||
| AK172238.1 | - | 2 - 3 | 1175 | 78 | ||
| BC036336.1 | - | 1 - 2 | 741 | 78 | ||
| BC064447.1 | - | 2 - 3 | 1187 | 78 | ||
More Information
Set Membership: |
|

Whsc1l1
Genomic Map