Assay Details
| Gene Symbol: Pan3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028291.4 | NP_082567.3 | 15 - 16 | 2154 | 113 | |
| GenBank mRNA | AK012409.1 | - | 4 - 5 | 550 | 113 | |
| BC061486.1 | - | 4 - 5 | 747 | 113 | ||
| BC082547.1 | - | 14 - 15 | 1683 | 6838444 | 113 | |
| BC086321.1 | - | 8 - 9 | 891 | 30108704 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pan3
Genomic Map