Assay Details
| Gene Symbol: Pan3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028291.4 | NP_082567.3 | 17 - 18 | 2363 | 88 | |
| GenBank mRNA | AK012409.1 | - | 6 - 7 | 759 | 88 | |
| AK028869.1 | - | 2 - 3 | 173 | 88 | ||
| BC013826.1 | - | 2 - 3 | 193 | 88 | ||
| BC082547.1 | - | 16 - 17 | 1892 | 6838444 | 88 | |
| BC086321.1 | - | 10 - 11 | 1100 | 30108704 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pan3
Genomic Map