Assay Details
Gene Symbol: Rxfp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289564.1 | NP_001276493.1 | 11 - 12 | 1070 | 67 | |
NM_001289566.1 | NP_001276495.1 | 11 - 12 | 1112 | 67 | ||
NM_080468.2 | NP_536716.2 | 12 - 13 | 1142 | 67 | ||
GenBank mRNA | AF346501.1 | - | 12 - 13 | 1142 | 67 | |
AK034592.1 | - | 12 - 13 | 1055 | 67 | ||
BC120741.1 | - | 12 - 13 | 1111 | 40129664 | 67 | |
BC144802.1 | - | 11 - 12 | 1039 | 9053350 | 67 | |
BC144803.1 | - | 11 - 12 | 1081 | 9053351 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
