Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500017087.mm.1 |
TC0500001859.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500017087.mm.1 |
NM_001289566.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500017086.mm.1 |
TC0500001859.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500017086.mm.1 |
NM_001289564.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500017086.mm.1 |
NM_001289566.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500017086.mm.1 |
NM_080468.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500017086.mm.1 |
NM_080468.2 |
Refseq |