Assay Details
Gene Symbol: Rxfp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289564.1 | NP_001276493.1 | 8 - 9 | 857 | 79 | |
NM_001289566.1 | NP_001276495.1 | 8 - 9 | 899 | 79 | ||
NM_080468.2 | NP_536716.2 | 8 - 9 | 857 | 79 | ||
GenBank mRNA | AK034592.1 | - | 8 - 9 | 770 | 79 | |
BC120741.1 | - | 8 - 9 | 826 | 40129664 | 79 | |
BC144802.1 | - | 8 - 9 | 826 | 9053350 | 79 | |
BC144803.1 | - | 8 - 9 | 868 | 9053351 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
