| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400018302.mm.1 |
NR_045185.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018302.mm.1 |
NR_045185.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018302.mm.1 |
TC0400002194.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400018313.mm.1 |
NR_045185.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018313.mm.1 |
NR_045185.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018313.mm.1 |
TC0400002194.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400018305.mm.1 |
NR_045185.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018305.mm.1 |
NR_045185.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018305.mm.1 |
TC0400002194.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400018311.mm.1 |
NR_045185.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018311.mm.1 |
NR_045185.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018311.mm.1 |
TC0400002194.mm.1 |
MTA TC |