Assay Details
| Gene Symbol: Alox5ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004002.1 | - | 3 - 4 | 305 | 77 | |
| AK075967.1 | - | 3 - 4 | 288 | 77 | ||
| BC026209.1 | - | 3 - 4 | 292 | 1349819 | 77 | |
| M96554.1 | - | 3 - 4 | 237 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alox5ap
Genomic Map