Assay Details
| Gene Symbol: Hsph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013559.2 | NP_038587.2 | 11 - 12 | 1816 | 66 | |
| GenBank mRNA | AK075731.1 | - | 1 - 2 | 39 | 66 | |
| AK083179.1 | - | 1677 | 66 | |||
| AK146697.1 | - | 11 - 12 | 1735 | 66 | ||
| AK160824.1 | - | 11 - 12 | 1797 | 66 | ||
| AK165046.1 | - | 11 - 12 | 1784 | 66 | ||
| AK172913.1 | - | 5 - 6 | 713 | 66 | ||
| BC018378.1 | - | 11 - 12 | 1691 | 3257042 | 66 | |
| D67016.1 | - | 11 - 12 | 1631 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsph1
Genomic Map