Assay Details
| Gene Symbol: Hsph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013559.2 | NP_038587.2 | 2 - 3 | 406 | 117 | |
| GenBank mRNA | AK083179.1 | - | 267 | 117 | ||
| AK146697.1 | - | 2 - 3 | 325 | 117 | ||
| AK160824.1 | - | 2 - 3 | 387 | 117 | ||
| AK165046.1 | - | 2 - 3 | 374 | 117 | ||
| BC018378.1 | - | 2 - 3 | 281 | 3257042 | 117 | |
| CF164049.1 | - | 2 - 3 | 406 | 117 | ||
| D67016.1 | - | 2 - 3 | 221 | 117 | ||
| D67017.1 | - | 2 - 3 | 308 | 117 | ||
| L40406.1 | - | 2 - 3 | 306 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsph1
Genomic Map