Assay Details
| Gene Symbol: Hsph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013559.2 | NP_038587.2 | 3 - 4 | 545 | 89 | |
| GenBank mRNA | AK083179.1 | - | 406 | 89 | ||
| AK146697.1 | - | 3 - 4 | 464 | 89 | ||
| AK160824.1 | - | 3 - 4 | 526 | 89 | ||
| AK165046.1 | - | 3 - 4 | 513 | 89 | ||
| BC018378.1 | - | 3 - 4 | 420 | 3257042 | 89 | |
| D67016.1 | - | 3 - 4 | 360 | 89 | ||
| D67017.1 | - | 3 - 4 | 447 | 89 | ||
| L40406.1 | - | 3 - 4 | 445 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsph1
Genomic Map