Assay Details
| Gene Symbol: Gm15408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040429.1 | 1 - 2 | 333 | 79 | ||
| GenBank mRNA | AK087624.1 | - | 1 - 2 | 65 | 79 | |
| CJ159174.1 | - | 1 - 2 | 334 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 15411 | Gm15411 | OTTMUSG00000021229 | Anti-sense | Chr. 5 - 148953741 - 149015813 on NCBI Build GRCm38 | |
| Downstream | high mobility group box 1 | Hmgb1 |
DEF, HMG-1, Hmg1, SBP-1, amphoterin, p30 |
39.2kb | Anti-sense | Chr. 5 - 149047222 - 149053037 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm15408
Genomic Map